Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000758.3 | NP_000749.2 | 3 - 4 | 359 | 121 | |
| GenBank mRNA | AY720441.1 | - | 3 - 4 | 336 | 121 | |
| BC108724.1 | - | 3 - 4 | 346 | 3886571 | 121 | |
| BC113924.1 | - | 3 - 4 | 350 | 40084800 | 121 | |
| BC113999.1 | - | 3 - 4 | 350 | 40084798 | 121 | |
| KP271009.1 | - | 3 - 4 | 276 | 121 | ||
| M10663.1 | - | 3 - 4 | 335 | 121 | ||
| M11220.1 | - | 3 - 4 | 359 | 121 | ||
| M11734.1 | - | 3 - 4 | 342 | 121 | ||
More Information
Set Membership: |
|

CSF2
Genomic Map