Assay Details
| Gene Symbol: CDKN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058195.3 | NP_478102.2 | 1 - 2 | 350 | 72 | |
| GenBank mRNA | BC015960.2 | - | 1 - 2 | 240 | 72 | |
| BC021998.2 | - | 1 - 2 | 240 | 72 | ||
| BT007020.1 | - | 1 - 2 | 190 | 72 | ||
| S78535.1 | - | 1 - 2 | 331 | 72 | ||
| U26727.1 | - | 1 - 2 | 217 | 72 | ||
| U38945.1 | - | 1 - 2 | 350 | 72 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Citations (1)
Clinical and biological significance of CDK4 amplification in well-differentiated and dedifferentiated liposarcomas.
- Authors
- Italiano A, Bianchini L, Gjernes E, Keslair F, Ranchere-Vince D, Dumollard JM, Haudebourg J, Leroux A, Mainguene C, Terrier P, Chibon F, Coindre JM, Pedeutour F
- Journal
- Clin Cancer Res - Page No 5696 (Tue Sep 15 00:00:00 UTC 2009)
- Volume (Year)
- 15 (2009)
- Product Usage
- -

CDKN2A
Genomic Map