Assay Details
| Gene Symbol: RASSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576736.2 | XP_005576793.1 | 2 - 3 | 218 | 71 | |
| XM_015434209.1 | XP_015289695.1 | 2 - 3 | 218 | 71 | ||
| XM_015434211.1 | XP_015289697.1 | 2 - 3 | 218 | 71 | ||
More Information
Set Membership: |
|

RASSF7
Genomic Map