Assay Details
| Gene Symbol: HSD17B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571262.2 | XP_005571319.1 | 3 - 4 | 757 | 84 | |
| XM_005571263.2 | XP_005571320.1 | 3 - 4 | 781 | 84 | ||
| XM_005571264.2 | XP_005571321.1 | 3 - 4 | 677 | 84 | ||
| XM_005571265.2 | XP_005571322.1 | 3 - 4 | 758 | 84 | ||
| GenBank mRNA | AB171697.1 | - | 3 - 4 | 704 | 84 | |
More Information
Set Membership: |
|

HSD17B6
Genomic Map