Assay Details
| Gene Symbol: ZCCHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285353.1 | NP_001272282.1 | 5 - 6 | 788 | 79 | |
| XM_005557294.2 | XP_005557351.1 | 5 - 6 | 824 | 79 | ||
| GenBank mRNA | AB174564.1 | - | 5 - 6 | 788 | 79 | |
More Information
Set Membership: |
|

ZCCHC9
Genomic Map