Assay Details
| Gene Symbol: LSM14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588775.2 | XP_005588832.1 | 1 - 2 | 318 | 74 | |
| XM_005588776.2 | XP_005588833.1 | 1 - 2 | 318 | 74 | ||
| XM_015441557.1 | XP_015297043.1 | 1 - 2 | 301 | 74 | ||
| XM_015441558.1 | XP_015297044.1 | 1 - 2 | 301 | 74 | ||
More Information
Set Membership: |
|

LSM14A
Genomic Map