Assay Details
| Gene Symbol: IRF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283529.1 | NP_001270458.1 | 6 - 7 | 692 | 67 | |
| XM_005560936.2 | XP_005560993.1 | 6 - 7 | 813 | 67 | ||
| GenBank mRNA | AB171112.1 | - | 6 - 7 | 692 | 67 | |
More Information
Set Membership: |
|

IRF9
Genomic Map