Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284619.1 | NP_001271548.1 | 8 - 9 | 1749 | 78 | |
| XM_005581402.2 | XP_005581459.1 | 8 - 9 | 1838 | 78 | ||
| XM_005581403.2 | XP_005581460.1 | 8 - 9 | 1979 | 78 | ||
| XM_005581404.1 | XP_005581461.1 | 8 - 9 | 1813 | 78 | ||
| XM_005581405.2 | XP_005581462.1 | 8 - 9 | 2348 | 78 | ||
| XM_005581407.2 | XP_005581464.1 | 8 - 9 | 1772 | 78 | ||
| XM_005581410.2 | XP_005581467.1 | 9 - 10 | 1724 | 78 | ||
| XM_015436502.1 | XP_015291988.1 | 8 - 9 | 1736 | 78 | ||
| GenBank mRNA | AB169313.1 | - | 8 - 9 | 1749 | 78 | |
More Information
Set Membership: |
|

FAM214B
Genomic Map