Assay Details
| Gene Symbol: FA2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283683.1 | NP_001270612.1 | 5 - 6 | 854 | 62 | |
| GenBank mRNA | AB169850.1 | - | 5 - 6 | 854 | 62 | |
More Information
Set Membership: |
|

FA2H
Genomic Map