Assay Details
| Gene Symbol: SLC35E2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545051.2 | XP_005545108.1 | 3 - 4 | 797 | 70 | |
| XM_015443854.1 | XP_015299340.1 | 3 - 4 | 797 | 70 | ||
| XM_015443861.1 | XP_015299347.1 | 3 - 4 | 797 | 70 | ||
More Information
Set Membership: |
|

SLC35E2B
Genomic Map