Assay Details
| Gene Symbol: C16H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583403.2 | XP_005583460.1 | 9 - 10 | 1027 | 82 | |
| GenBank mRNA | AB170397.1 | - | 9 - 10 | 1010 | 82 | |
More Information
Set Membership: |
|

C16H17orf75
Genomic Map