Assay Details
| Gene Symbol: EFNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541611.2 | XP_005541668.1 | 1 - 2 | 237 | 98 | |
| XM_005541612.2 | XP_005541669.1 | 1 - 2 | 237 | 98 | ||
More Information
Set Membership: |
|

EFNA3
Genomic Map