Assay Details
| Gene Symbol: IL23A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287659.1 | NP_001274588.1 | 3 - 4 | 489 | 82 | |
| XM_005571219.2 | XP_005571276.1 | 4 - 5 | 576 | 82 | ||
| GenBank mRNA | JQ288993.1 | - | 3 - 4 | 489 | 82 | |
More Information
Set Membership: |
|

IL23A
Genomic Map