Assay Details
| Gene Symbol: SESN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284868.1 | NP_001271797.1 | 2 - 3 | 1088 | 60 | |
| XM_015448373.1 | XP_015303859.1 | 3 - 4 | 981 | 60 | ||
| XM_015448374.1 | XP_015303860.1 | 3 - 4 | 986 | 60 | ||
| XM_015448375.1 | XP_015303861.1 | 2 - 3 | 251 | 60 | ||
| XM_015448376.1 | XP_015303862.1 | 2 - 3 | 225 | 60 | ||
| GenBank mRNA | AB168675.1 | - | 2 - 3 | 1088 | 60 | |
| AB169134.1 | - | 3 - 4 | 282 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015448376.1 transcripts.
Set Membership: |
|

SESN1
Genomic Map