Assay Details
| Gene Symbol: LOC102133073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581112.2 | XP_005581169.2 | 2 - 3 | 120 | 69 | |
| XM_015436868.1 | XP_015292354.1 | 2 - 3 | 112 | 69 | ||
| XM_015436869.1 | XP_015292355.1 | 2 - 3 | 112 | 69 | ||
| XM_015436870.1 | XP_015292356.1 | 2 - 3 | 112 | 69 | ||
| XM_015436871.1 | XP_015292357.1 | 2 - 3 | 112 | 69 | ||
| XM_015436873.1 | XP_015292359.1 | 3 - 4 | 362 | 69 | ||
More Information
Set Membership: |
|

LOC102133073
Genomic Map