Assay Details
| Gene Symbol: RBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283094.1 | NP_001270023.1 | 1 - 2 | 93 | 82 | |
| GenBank mRNA | AB170446.1 | - | 1 - 2 | 93 | 82 | |
| AB220470.1 | - | 1 - 2 | 93 | 82 | ||
More Information
Set Membership: |
|

RBX1
Genomic Map