Assay Details
| Gene Symbol: QKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448508.1 | XP_015303994.1 | 6 - 7 | 2287 | 82 | |
| XM_015448510.1 | XP_015303996.1 | 6 - 7 | 2407 | 82 | ||
| XM_015448511.1 | XP_015303997.1 | 6 - 7 | 2379 | 82 | ||
More Information
Set Membership: |
|

QKI
Genomic Map