Assay Details
| Gene Symbol: E2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544468.2 | XP_005544525.1 | 4 - 5 | 1285 | 64 | |
| XM_005544469.2 | XP_005544526.1 | 4 - 5 | 1279 | 64 | ||
More Information
Set Membership: |
|

E2F2
Genomic Map