Assay Details
| Gene Symbol: WASHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564056.2 | XP_005564113.1 | 5 - 6 | 846 | 99 | |
| XM_015455165.1 | XP_015310651.1 | 6 - 7 | 947 | 99 | ||
More Information
Set Membership: |
|

WASHC5
Genomic Map