Assay Details
| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283254.1 | NP_001270183.1 | 3 - 4 | 502 | 102 | |
| XM_005547681.2 | XP_005547738.1 | 3 - 4 | 431 | 102 | ||
| GenBank mRNA | AB179484.1 | - | 3 - 4 | 503 | 102 | |
More Information
Set Membership: |
|

LMCD1
Genomic Map