Assay Details
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594059.2 | XP_005594116.1 | 9 - 10 | 1681 | 274 | |
| XM_005594061.2 | XP_005594118.1 | 8 - 9 | 1543 | 136 | ||
| XM_005594062.2 | XP_005594119.1 | 7 - 8 | 1289 | 136 | ||
| XM_015443874.1 | XP_015299360.1 | 6 - 7 | 2636 | 274 | ||
| XM_015443875.1 | XP_015299361.1 | 9 - 10 | 1686 | 274 | ||
| XM_015443876.1 | XP_015299362.1 | 9 - 10 | 1570 | 274 | ||
| XM_015443877.1 | XP_015299363.1 | 8 - 9 | 1427 | 274 | ||
More Information
Set Membership: |
|

ZNF711
Genomic Map