Assay Details
| Gene Symbol: RBM38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569422.2 | XP_005569479.1 | 1 - 2 | 221 | 79 | |
| XM_005569423.2 | XP_005569480.1 | 1 - 2 | 221 | 79 | ||
More Information
Set Membership: |
|

RBM38
Genomic Map