Assay Details
| Gene Symbol: LRRC17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447873.1 | XP_015303359.1 | 1 - 2 | 826 | 140 | |
| XM_015447875.1 | XP_015303361.1 | 1 - 2 | 826 | 140 | ||
More Information
Set Membership: |
|

LRRC17
Genomic Map