Assay Details
| Gene Symbol: UBE2Q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560135.2 | XP_005560192.1 | 11 - 12 | 1398 | 75 | |
| XM_005560136.2 | XP_005560193.1 | 11 - 12 | 1273 | 75 | ||
| XM_005560137.2 | XP_005560194.1 | 10 - 11 | 1293 | 75 | ||
| XM_005560138.2 | XP_005560195.1 | 8 - 9 | 1161 | 75 | ||
More Information
Set Membership: |
|

UBE2Q2
Genomic Map