Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283557.1 | NP_001270486.1 | 2 - 3 | 987 | 73 | |
| XM_005556783.2 | XP_005556840.1 | 2 - 3 | 2013 | 73 | ||
| GenBank mRNA | AB169243.1 | - | 2 - 3 | 987 | 73 | |
| AY052643.1 | - | 1 - 2 | 870 | 73 | ||
More Information
Set Membership: |
|

PTGER4
Genomic Map