Assay Details
| Gene Symbol: MIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283697.1 | NP_001270626.1 | 392 | 56 | ||
| GenBank mRNA | AB169695.1 | - | 392 | 56 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIF| Gene Symbol: MIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283697.1 | NP_001270626.1 | 392 | 56 | ||
| GenBank mRNA | AB169695.1 | - | 392 | 56 | ||
Set Membership: |
|