Assay Details
| Gene Symbol: SUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285113.1 | NP_001272042.1 | 4 - 5 | 355 | 75 | |
| XM_015451155.1 | XP_015306641.1 | 4 - 5 | 393 | 75 | ||
| XM_015451156.1 | XP_015306642.1 | 4 - 5 | 322 | 75 | ||
| GenBank mRNA | AB168249.1 | - | 4 - 5 | 355 | 75 | |
| AB170806.1 | - | 4 - 5 | 372 | 75 | ||
| AB174427.1 | - | 4 - 5 | 355 | 75 | ||
More Information
Set Membership: |
|

SUB1
Genomic Map