Assay Details
| Gene Symbol: NHLH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430612.1 | XP_015286098.1 | 1 - 2 | 582 | 79 | |
| XM_015430613.1 | XP_015286099.1 | 1 - 2 | 582 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015430612.1, XM_015430613.1 transcripts.
Set Membership: |
|

NHLH2
Genomic Map