Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283911.1 | NP_001270840.1 | 8 - 9 | 1089 | 99 | |
| GenBank mRNA | AB171327.1 | - | 8 - 9 | 1089 | 99 | |
More Information
Set Membership: |
|

ZC3H15
Genomic Map