Assay Details
| Gene Symbol: LCMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283175.1 | NP_001270104.1 | 5 - 6 | 597 | 84 | |
| XM_005591495.2 | XP_005591552.1 | 5 - 6 | 622 | 84 | ||
| XM_005591498.2 | XP_005591555.1 | 5 - 6 | 622 | 84 | ||
| XM_015442158.1 | XP_015297644.1 | 5 - 6 | 622 | 84 | ||
| XM_015442159.1 | XP_015297645.1 | 5 - 6 | 622 | 84 | ||
| XM_015442160.1 | XP_015297646.1 | 5 - 6 | 656 | 84 | ||
| XM_015442161.1 | XP_015297647.1 | 5 - 6 | 622 | 84 | ||
| GenBank mRNA | AB171364.1 | - | 5 - 6 | 597 | 84 | |
More Information
Set Membership: |
|

LCMT1
Genomic Map