Assay Details
| Gene Symbol: MOB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543475.2 | XP_005543532.1 | 3 - 4 | 850 | 85 | |
| XM_015436442.1 | XP_015291928.1 | 2 - 3 | 3686 | 85 | ||
More Information
Set Membership: |
|

MOB3C
Genomic Map