Assay Details
| Gene Symbol: PPP6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283810.1 | NP_001270739.1 | 4 - 5 | 544 | 83 | |
| GenBank mRNA | AB172685.1 | - | 4 - 5 | 544 | 83 | |
More Information
Set Membership: |
|

PPP6C
Genomic Map