Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449721.1 | XP_015305207.1 | 2 - 3 | 1382 | 89 | |
| XM_015449722.1 | XP_015305208.1 | 3 - 4 | 653 | 89 | ||
More Information
Set Membership: |
|

ZSCAN16
Genomic Map