Assay Details
| Gene Symbol: ARF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283926.1 | NP_001270855.1 | 2 - 3 | 197 | 114 | |
| XM_015447255.1 | XP_015302741.1 | 2 - 3 | 287 | 114 | ||
| GenBank mRNA | AB170135.1 | - | 2 - 3 | 215 | 114 | |
| AB171125.1 | - | 2 - 3 | 197 | 114 | ||
More Information
Set Membership: |
|

ARF5
Genomic Map