Assay Details
| Gene Symbol: EIF2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283137.1 | NP_001270066.1 | 5 - 6 | 1419 | 81 | |
| XM_015453235.1 | XP_015308721.1 | 6 - 7 | 890 | 81 | ||
| GenBank mRNA | AB172712.1 | - | 5 - 6 | 1419 | 81 | |
| AY650291.1 | - | 4 - 5 | 428 | 81 | ||
More Information
Set Membership: |
|

EIF2S1
Genomic Map