Assay Details
| Gene Symbol: PPP1CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576233.2 | XP_005576290.1 | 2 - 3 | 248 | 62 | |
| XM_015433838.1 | XP_015289324.1 | 2 - 3 | 237 | 62 | ||
More Information
Set Membership: |
|

PPP1CB
Genomic Map