Assay Details
| Gene Symbol: PPP1R9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583654.2 | XP_005583711.1 | 5 - 6 | 2053 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R9B| Gene Symbol: PPP1R9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583654.2 | XP_005583711.1 | 5 - 6 | 2053 | 52 | |
Set Membership: |
|