Assay Details
| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543607.2 | XP_005543664.1 | 2 - 3 | 531 | 65 | |
| XM_005543608.2 | XP_005543665.1 | 2 - 3 | 492 | 65 | ||
More Information
Set Membership: |
|

B4GALT2
Genomic Map