Assay Details
| Gene Symbol: YDJC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567874.2 | XP_005567931.1 | 1 - 2 | 423 | 101 | |
| XM_005567875.2 | XP_005567932.1 | 1 - 2 | 423 | 101 | ||
More Information
Set Membership: |
|

YDJC
Genomic Map