Assay Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554819.2 | XP_005554876.1 | 1 - 2 | 143 | 77 | |
| XM_005554820.2 | XP_005554877.1 | 1 - 2 | 143 | 77 | ||
| XM_005554822.2 | XP_005554879.1 | 1 - 2 | 143 | 77 | ||
More Information
Set Membership: |
|

COMMD8
Genomic Map