Assay Details
| Gene Symbol: SLC36A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283600.1 | NP_001270529.1 | 10 - 11 | 1293 | 101 | |
| XM_005558314.2 | XP_005558371.1 | 10 - 11 | 1380 | 101 | ||
| XM_005558316.2 | XP_005558373.1 | 10 - 11 | 1167 | 101 | ||
| XM_015451355.1 | XP_015306841.1 | 10 - 11 | 1431 | 101 | ||
| GenBank mRNA | AB168788.1 | - | 10 - 11 | 1293 | 101 | |
More Information
Set Membership: |
|

SLC36A1
Genomic Map