Assay Details
| Gene Symbol: LOC102124147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590006.2 | XP_005590063.1 | 9 - 10 | 1424 | 69 | |
| XM_015441358.1 | XP_015296844.1 | 9 - 10 | 1425 | 69 | ||
More Information
Set Membership: |
|

LOC102124147
Genomic Map