Assay Details
| Gene Symbol: BCL11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562182.2 | XP_005562239.1 | 2 - 3 | 1437 | 66 | |
| XM_005562183.2 | XP_005562240.1 | 2 - 3 | 1434 | 66 | ||
More Information
Set Membership: |
|

BCL11B
Genomic Map