Assay Details
| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285032.1 | NP_001271961.1 | 6 - 7 | 1341 | 61 | |
| GenBank mRNA | AB171522.1 | - | 6 - 7 | 1341 | 61 | |
More Information
Set Membership: |
|

SNTB2
Genomic Map