Assay Details
| Gene Symbol: NKX2-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457737.1 | XP_015313223.1 | 1 - 2 | 8440 | 147 | |
| GenBank mRNA | AY775948.1 | - | 1 - 2 | 96 | 147 | |
More Information
Set Membership: |
|

NKX2-2
Genomic Map