Assay Details
| Gene Symbol: NOSIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284556.1 | NP_001271485.1 | 2 - 3 | 238 | 56 | |
| GenBank mRNA | AB168844.1 | - | 2 - 3 | 238 | 56 | |
More Information
Set Membership: |
|

NOSIP
Genomic Map