Assay Details
| Gene Symbol: CYP3A43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319505.1 | NP_001306434.1 | 11 - 12 | 1332 | 99 | |
| XM_005549171.2 | XP_005549228.1 | 10 - 11 | 1520 | 99 | ||
| XM_015447122.1 | XP_015302608.1 | 11 - 12 | 1614 | 99 | ||
| XM_015447123.1 | XP_015302609.1 | 10 - 11 | 1514 | 99 | ||
| XM_015447124.1 | XP_015302610.1 | 9 - 10 | 1420 | 99 | ||
| XM_015447125.1 | XP_015302611.1 | 8 - 9 | 1003 | 99 | ||
| GenBank mRNA | DQ074796.1 | - | 11 - 12 | 1332 | 99 | |
More Information
Set Membership: |
|

CYP3A43
Genomic Map