Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319571.1 | NP_001306500.1 | 7 - 8 | 980 | 90 | |
| XM_005593641.2 | XP_005593698.1 | 7 - 8 | 1099 | 90 | ||
| XM_005593642.2 | XP_005593699.1 | 7 - 8 | 953 | 90 | ||
| XM_005593643.2 | XP_005593700.1 | 6 - 7 | 884 | 90 | ||
| XM_005593644.1 | XP_005593701.1 | 6 - 7 | 630 | 90 | ||
| XM_015443501.1 | XP_015298987.1 | 7 - 8 | 947 | 90 | ||
| XM_015443502.1 | XP_015298988.1 | 6 - 7 | 1008 | 90 | ||
| XM_015443503.1 | XP_015298989.1 | 6 - 7 | 856 | 90 | ||
| GenBank mRNA | AB169289.1 | - | 7 - 8 | 980 | 90 | |
More Information
Set Membership: |
|

RIBC1
Genomic Map