Assay Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578650.2 | XP_005578707.1 | 2 - 3 | 567 | 110 | |
| XM_005578651.1 | XP_005578708.1 | 2 - 3 | 150 | 110 | ||
More Information
Set Membership: |
|

TMEM41B
Genomic Map