Assay Details
| Gene Symbol: GNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550379.2 | XP_005550436.1 | 5 - 6 | 6139 | 92 | |
| XM_015447849.1 | XP_015303335.1 | 5 - 6 | 898 | 92 | ||
More Information
Set Membership: |
|

GNAT3
Genomic Map